Environmental Express® Digestion Tubes and Accessories
Digestion Cups and Accessories
Shop Environmental Express® Digestion Tubes >
Our polypropylene, screw-cap digestion vessels come in a variety of sizes to fit each of our HotBlocks. These convenient, disposable tubes eliminate the need for glassware and associated cleanup. Vessels are certified for low metals background and do not require acid washing prior to use. All digestion vessels have molded in graduations and serve as graduated cylinders as well as sample storage containers. After the digestion procedure is complete, samples can be brought to volume and capped for storage, eliminating transfer steps. All digestion vessels have molded in lift rings to ensure secure positioning in sample transfer racks.
If you need assistance, please do not hesitate to call us at 1-800-343-5319.
Method Compliance: This product complies with the following methods:
EPA 200.2 EPA 200.5 EPA 200.7
EPA 200.8 EPA 200.9 EPA 245.1
EPA 245.7 SM 3030 SM 3112
SM 3120 SM 3125 ISO 15202-2:2012
ISO 15587-1:2002 ISO 15587-2:2002 EPA 3050B
EPA 7470 EPA 7471  
Don't see your test method listed? Please let us know. You can email us at [email protected]