Max Flow Rate (GPM)
Max Flow Rate (LPM)
Max Pressure (bar)
Power (VAC)
Chemsteel
Chemsteel
Chemsteel provides a variety of gear pumps that range in flow rates from 1.5 GPM to 30 GPM. These pumps are self-priming and bidirectional for dosing, metering, and transferring of polymers and corrosive chemicals. We are a Chemsteel authorized distributo