Availability
Max Capacity (mL)
Power (VAC)
CINC
CINC
CINC separator/extractors mix, react, and/or separate liquid-liquid mixtures in a single device. Their design operates effectively under changing inlet concentrations, variable flow rates, and flow disruptions. We are a CINC authorized distributor.