Availability
Color
Material
Size
Dunlop
Dunlop
Dunlop® Protective Footwear is a leading global provider of protective footwear worn by workers in 50 countries. Continuous innovation and handcrafting makes this footwear safe and comfortable. Some of the products are manufactured with Purofort®, the unique Dunlop material with millions of evenly distributed air pockets making the footwear lightweight and thermally insulating, plus a cross-lined structure that provides flexibility and strength. Dunlop Protective Footwear is ideal for workers in agriculture, fishery, food processing, the oil, gas & mining industries, and anyone wanting footwear that not only protects but is also comfortable to wear for long periods of time.