GE Panametrics
GE Panametrics
GE Panametrics ultrasonic flowmeter kits measure flow from outside plastic, metal, or concrete-lined pipes to prevent pressure drop, leaks, or contamination. Select from kits for various pipe sizes. We are a GE Panametrics authorized distributor.