Availability
Max Flow Rate (GPM)
Max Flow Rate (LPM)
Power (VAC)
Seepex
Seepex
seepex manufactures progressing cavity pumps, offered here in two configurations: iron bodies with NBR stators and stainless steel (SS) bodies with Viton® stators. Pumps are adaptable to a wide range of industries. We are a seepex authorized distributor.