An economical new video borescope inspection camera takes users where they haven’t been before. The miniature waterproof camera head allows high-resolution viewing in small spaces—ideal for automotive applications, building maintenance, and laboratory analysis.
The Extech Video Borescope Inspection Camera features a 17-mm diameter camera head with four built-in illuminating LEDs and a 2.4″ color TFT LCD monitor. Video can be output for viewing on any monitor with a video input jack. Images are glare-free even with close-up viewing. A 39″L flexible gooseneck cable retains its configured shape.
A wireless version is also available with a 9-mm diameter camera head and a detachable 3.5″ color TFT LCD wireless monitor with 2GB microSD memory card for capturing images (JPEG) or video (AVI) for viewing on your PC. The wireless LCD can be viewed from a remote location up to 32 feet from measurement point. Its menu displays 10 languages.
Be the first to comment on "Innovative New Cameras Offer Views into Cramped Spaces―for Inspection and Analysis"