pH is a fundamental element of the wine-making industry. pH strongly influences wine properties such as color, oxidation, biological and chemical stability. A meter with up to 3-point pH calibration or a handheld meter with 5-point pH calibration are accurate and reliable tools for measuring pH levels in wine.
pH measures the quantity of acids present, the strength of the acids, and the effects of minerals and other ingredients in the wine. Wine pH depends on three main factors: the total amount of acid present, the ratio of malic acid to tartaric acid, and the amount of potassium present. Wines that contain little acid and excess potassium show high pH values. Wine with more tartaric acid, less malic acid, less potassium, and more titratable acid has lower pH values.
pH values range from 2.9 to 4.2 in wine. Wine’s chemical and biological stability are very dependent on pH value. Lower pH values are known to improve the stability, so winemakers usually prefer a pH range of 3.0 to 3.5. The wine is so stable in this range that many winemakers believe pH is a crucial guideline in wine-making.
There are many advantages to low pH values in wine. Low pH inhibits bacteria, causes sugar fermentation to progress more evenly, and makes malolactic fermentation easier to control. Low pH also has a direct influence on the hot stability of wine. When bottled wines are stored in warm areas, protein precipitates out of them, causing serious problems. These wines are then treated with bentonite, which removes excess protein. pH is important to the treatment because bentonite successfully removes more protein when the pH value is low. If wine pH increases, bentonite is less effective, making it necessary to add larger amounts. The danger is adding too much bentonite because it can strip wines of their unique aromas and flavors. Read more.
Be the first to comment on "Measuring pH in Wine-Making"