Understanding Your Lab Equipment: How a Refractometer Works
Refractometer
New Cole-Parmer® Digital Refractometer
Do you have a refractometer?
A refractometer is a simple instrument used for measuring concentrations of aqueous solutions. It requires only a few drops of liquid and is used as lab equipment throughout the food, agricultural, chemical, and manufacturing industries.
Refraction
When light enters a liquid it changes direction; this is called refraction. Refractometers measure the degree to which the light changes direction, called the angle of refraction. A refractometer takes the refraction angles and correlates them to refractive index (nD) values that have been established. Using these values, you can determine the concentrations of solutions. For example, solutions have different refractive indexes depending on their concentration.
The prism in the refractometer has a greater refractive index than the solution. Measurements are read at the point where the prism and solution meet. With a low concentration solution, the refractive index of the prism is much greater than that of the sample, creating a large refraction angle and a low reading. The reverse would happen with a high concentration solution.
Learn more about the new Cole-Parmer digital refractometers.
The % Brix Scale
The % Brix Scale shows the concentration percentage of the soluble solids content of a sample (water solution). The soluble solids content is the total of all the solids dissolved in the water, beginning with sugar, salts, protein, acids, etc.; the measurement reading value is the value of the sum total of those. The Brix scale is calibrated to the number of grams of cane sugar contained in 100 g of water. When measuring a sugar solution, the % Brix reading should perfectly match the actual concentration.
Sample fluid
% Brix
Cutting oils
0 to 8
Oranges
4 to 13
Carbonated beverages
5 to 15
Apples
11 to 18
Grapes and wines
14 to 19
Concentrated juices
42 to 68
Condensed milk
52 to 68
Jams and jellies
60 to 70
Common Refractive Indexes
Refractive index readings are temperature-dependent.
Sample fluid
Temperature
Refractive index
Methanol
25°C
1.326
Acetone
25°C
1.357
Ethanol
25°C
1.359
Acetic acid
25°C
1.370
Benzene
25°C
1.498
Paraffin oil
20°C
1.412
Palm oil
20°C
1.456
Olive oil
20°C
1.471
Methyl salicylate
25°C
1.522
Methyl iodide
25°C
1.740
Need to know more about refractometers? Read the entire article on refractometers.
27 Comments on "Understanding Your Lab Equipment: How a Refractometer Works"
  1. Tariro Linda Majakire | March 28, 2017 at 3:28 am | Reply
    what is the range for a perfect refractometer for palm oil
    • Thank you for your inquiry. Your question was forwarded to our technical product experts. You can also call 1-800-323-4340 for immediate assistance.
  2. licensed Sandra | June 20, 2017 at 4:59 am | Reply
    Way cool.  Love it!
  3. Masturina Suhaimi | May 19, 2019 at 8:58 pm | Reply
    what is the range for a perfect refractometer for honey ?
    • Thank you for your question. We will have someone from our applications department contact you. If you need an immediate answer, please call 1-800-323-4340.
  4. Oscar Alicea | June 5, 2019 at 8:34 am | Reply
    For application of large decal onto painted plastic we use a soap and water solution. This solution is measured out according to the established recipe. I would like to know if a refractometer can be used to verify the correct percentage of this solution is maintained in bulk container. Also could you tell me what would be the range for a soap and water solution. Thank You….
    • Hi Oscar:
      Thank you for your question. I forwarded your information to our application specialists. Someone will be in contact with you. In the meantime, if you need immediate assistance, please call 1-800-434-4340.
  5. will a refractometer measure moisture in molasses?
    Does a conversion chart exists that converts refractive index to moisture?
    Can you recommend a refractometer for this application?
    • Hi Tom:
      Thanks for reading our blog. Your questions have been forwarded to our customer service dept. In the meantime, if you need immediate assistance, please feel free to call 1-800-323-4340.
  6. Ajay choudhari | August 1, 2019 at 10:45 pm | Reply
    Near by mumbai in which lab refractometer ke test kr ke de skte he
    • Hi Ajay:
      Thank you for reaching out. I will send this to our team in India, and you should receive a response from them soon.
  7. Are refractometers used in gemology to identify gemstone and mineral samples?
    • Hello:
      We will send your question to our application experts so you can get a complete answer. Should you want immediate assistance, please call us at 1-800-323-4340.
    • Hello:
      There are special refractometers for gemology. Unfortunately, we do not sell them.
  8. I need this for a project pls | August 22, 2019 at 7:58 am | Reply
    Hey there, what’s the correct term used to call the part of the refractometer where the liquid sample is dripped on?
    • Dr. Julia Lock | August 28, 2019 at 9:11 am | Reply
      Thank you for your question. I will send it to our technical experts and they will get back to you. In the meantime, if you need it immediately, please feel free to call 1-800-323-4340.
  9. what is correlation between die coat concentration & Brix reading .
    • Hello:
      Thank you for your question. I’ve forwarded it to our technical experts and someone will email you.
    • Hi Rajkumar:
      Thank you for your question. I forwarded it to our technical representatives and someone will get back to you.
      Karen
  10. When measuring propylene glycol % in a mixture. What does the the +(-) number next to the % indicate?
    • Hello:
      The +(-) indicates the confidence interval or error in a measurement, often the standard deviation or standard error. If you have further questions, please reach out to our technical experts at 1-800-323-4340.
  11. Hey, i have a question if my sugar weight is 1500kg and i want to maintain 53 brix then how should i know what amount of water i should add to make it?
    • Cole-Parmer Blog Team | November 2, 2020 at 3:22 pm | Reply
      Hello Ibrar:
      Your inquiry was sent to our technical experts. You should hear from someone shortly. Thanks!
  12. Hi,
    I need to measure the concentration of peroxide to silane can I use a refractometer?
    kind regards,
    Alex
    • Cole-Parmer Blog Team | March 19, 2021 at 9:48 am | Reply
      Hi Alex:
      We sent your question to our technical experts. Someone will reach out to you. If you need an immediate answer, please call us at 1-800-323-4340. Thanks.
  13. Roberto Uribe | June 23, 2021 at 1:50 pm | Reply
    Is the refractometer can be use to check the concentration in dimethylpolysiloxane in a mix with water?
    • Antylia Scientific Blog Team | June 23, 2021 at 2:09 pm | Reply
      Hi Roberto:
      Thank you for your question. We will have one of our experts get back to you. If you need immediate assistance, please call 1-800-232-4340.
      Karen