Planning to attend WEFTEC (Water Environmental Federation Technical Exhibition and Conference) in Chicago? Stop by Cole-Parmer booth #3212 to see a selection of Oakton® meters. The conference, slated for October 7 to 9, is the largest water quality conference in North America.
Oakton Meters on display include:
Oakton® pH 5+ Meter is perfect for fast, accurate results. Automatic temperature compensation (ATC) ensures accuracy from 0 to 100°C. Meters offer a compact design with high +/-0.01 pH accuracy at an economical price.
Oakton® pH 700 Benchtop Meter is designed for today’s crowded laboratory benchtops. Its compact footprint is more than 40% smaller than other benchtop meters. This easy-to-use, microprocessor-based pH 700 benchtop meter measures pH, mV, relative mV, and temperature. The large, dual-display LCD provides excellent visibility—even from a distance.
Oakton® DO 2700 Benchtop Dissolved Oxygen Meter is an advanced meter for research grade use with BOD capability. It offers optimal performance and versatility and measures dissolved oxygen in mg/L, ppm, or % saturation with simultaneous temperature display in °F or °C.
Oakton® PTTestr 35 Waterproof Multiparameter pH/TDS Pocket Tester is the ultimate tester for pH, TDS, and temperature to combine the measurement capabilities of three testers in one convenient pocket-sized unit. Great for water, wastewater, laboratory or plant use, this multiparameter tester features long-life pH electrodes with a wide range of sample compatibility.
Also, meet Cole-Parmer Product Managers and Application Specialists at the booth to help you with any product or technical question. You can also register to win a DeWalt® Tool Set.
See all pH Meters and Water Quality Products.
Be the first to comment on "See Oakton Meters at WEFTEC"