See a special selection of lab equipment below that has been five-star rated by our customers!
“Very Nice” from a buyer in Arkansas
Cole-Parmer® Symmetry PA-Analytical Balance is an affordable option for precise weighing of small samples. It is built with advanced weighing technology and offers durability and easy-to-use operation. The balance features fast response times with the stabilization time approximately two seconds. It handles parts counting and percent weighing.
“Sampling done easier” from a customer in Florida
Masterflex® E/S™ Portable Sampler is an economical choice for basic applications. As part of the environmental sampling series from Masterflex, it is designed for portability and versatility. It features a single-turn potentiometer for speed control and three-position power/reversing switch. The sampler runs up to four hours on the self-contained 12 VDC rechargeable battery, or indefinitely from any AC or 12 VDC power supply. Charge the internal battery from an AC power supply or a DC voltage source of lower voltaic potential than the battery itself. The variable-speed drive has a top speed of 400 rpm, allowing you to adjust the flow rate to the needs of your application. The sampler floats up to 30 minutes if dropped in water when case is closed and latched.
“Excellent for the price” from a researcher in North Carolina
Furnaces with Three-Stage Programmable Control contain combined fiber and firebrick insulation that cools quickly and provides extended heating element life. A vertical lift door directs heat away from the user and also saves counter space. These furnaces provide maximum access with minimum head room for easy loading and unloading. Offer PID control with ten programs; a program link allows you to run two three-stage programs sequentially.
“Highly recommended” from a professional in Massachusetts
Storage Dewars (4 Liters) deliver superior vacuum performance. The vacuum space is filled with technologically advanced insulation for maximum holding times. The neck tube core minimizes liquid nitrogen losses. Optional roller base for easy moving of large capacity storage Dewars.
View our full selection of lab equipment.
Be the first to comment on "Scary Good Lab Equipment"