A Comparison of Metering Pumps (Dosing Pumps)
Metering pumps, also known as dosing pumps, are typically used when: high accuracy flow is required; the pumping is to be controlled by flow proportioning, computer, or PLC; flow rates are PLH or less; using high system pressure; and/or pumping high viscosity or slurry media. How do the various types of metering pumps compare? Here is an overview:
Bellows Pumps can pump gases or liquids and operate at flow rates from 6 mL/min to 2475 mL/min.
Metering Pump, Polypropylene Bellows
Metering Pump, Polypropylene Bellows
Maximum pressure: To 50 psi
Self-priming: Good
Pulseless flow: Poor
Viscous fluids: Fair
Particulate matter: Yes
Run dry: No
Diaphragm Pumps offer high accuracy for applications such as pH/ORP control. They run at flow rates from 10 mL/min to 19.558 L/min.
Maximum pressure: To 250 psi
Self-priming: Poor to Good
Pulseless flow: Poor
Viscous fluids: Good to excellent
Particulate matter: No
Run dry: Yes
Peristaltic Pumps are noncontaminating and available in a wide variety of pump materials. These tubing pumps operate at flow rates from 170 mL/min to 9.4 L/min.
Maximum pressure: To 125 psi
Self-priming: Excellent
Pulseless flow: Good at high rpm
Viscous fluids: Good
Particulate matter: Yes
Run dry: Yes
Polymer Feed Systems are used for polymer make down and run diluted polymer to 6.2 L/min.
Maximum pressure: To 100 psi
Self-priming: Poor
Pulseless flow: Poor
Viscous fluids: Excellent
Particulate matter: No
Run dry: N/A
Piston Pumps handle the highest pressure and deliver accuracy, which makes them ideal for HPLC applications. These pumps operate at flow rates of 0.016 mL/min to 404.6 L/min.
Maximum pressure: To 5000 psi
Self-priming: Good
Pulseless flow: Poor
Viscous fluids: Fair
Particulate matter: No
Run dry: No
Syringe Pumps operate with very low flow rates, from 0.001 µL/min to 147 mL/min.
Maximum pressure: To 40 psi
Self-priming: N/A
Pulseless flow: Excellent
Viscous fluids: Poor
Particulate matter: No
Run dry: Yes
Be the first to comment on "A Comparison of Metering Pumps (Dosing Pumps)"