Posted By: Cole-Parmer Blog Team
June 27, 2014
It’s quick and easy to load tubing into a Masterflex® Easy-Load® pump head! Just follow these two steps:
- Rotate lever to left to open head. Load tubing.
- Close head. Adjust side tubing retainers to hold tubing.
Like this:
Like Loading...
Related
Be the first to comment on "How to Load Tubing into Masterflex Easy-Load Pump Heads"