For research and development uses, Masterflex® tubing pump systems are an efficient choice. These peristaltic pumps are easy to load and versatile enough to use one pump drive for multiple applications. They offer exceptional repeatability for low volume dispensing and metering applications. Other benefits include:
- Non-contaminating because the fluid only contacts the inside of the tubing
- Low cell disruption—circulate cell tissue with minimal damage (low damage)
- Easy-to-clean by simply changing the tubing
- Multi-channel capabilities
- Mix/match pump heads, tubing and drives to suit applications—a selection of tubing is available that is compatible with numerous laboratory chemicals
Ten Typical R&D Applications for Masterflex Pumps:
- Acid/base dispensing
- Chromatography
- Desiccator air circulation
- Electrophoresis
- Filtration pumps
- Infusion pump for tissue
- Injection metering pump
- pH acid/base metering
- Spectrophotometer pump
- Toxicology assays (salt solutions). . .and more!
Be the first to comment on "10 R&D Applications that Benefit from Masterflex Pumps"