Syringe pumps are used in multi-channel, low flow, and pulseless applications. These highly accurate pumps are also referred to as infusion pumps.
Application Specialist Michael Hassan from our United Kingdom office shares his experienced knowledge of syringe pumps in this informational video.
Spend a few minutes with Michael as he takes you through the advantages and limitations of syringe pumps. Just choose Syringe Pump Overview on the Playlist.
Syringe pumps either draw in or push out liquid through a syringe to obtain a known volume as determined by the size of the syringe. You can depend on us for a wide selection of syringe pumps from low-flow, low-pressure applications, where precision is needed, to robust, high pressure models ideal for fluid delivery to reactors in chemical applications or for working with viscous fluids. Select pumps are available with user-friendly touch screens to help you work more efficiently.
Be the first to comment on "How do Syringe Pumps Work? Watch Video"