Looking for a laboratory furnace? Several variables to consider include process temperature, control requirements, pressure range, dimensions of samples and inner dimensions of the furnace chamber or tube.
Here are four types of laboratory furnaces that will help you narrow your choice:
- Muffle furnaces are your choice for high temperature uses such as ignition tests, gravimetric analysis, heat treating steel parts, and more
- Ashing furnaces can be ideal for determining the amount of ash in distillate and residual fuels, gas turbine fuels, crude oils, lubricating oils, waxes, and other petroleum products
- Tube furnaces are great for educational, governmental, and industrial laboratories for purifications and syntheses
- Melting furnaces safely melt silver, gold, brass and other metals without the potential hazards of torch melting or gas furnaces
View our selection of laboratory furnaces.
Be the first to comment on "4 Types of Laboratory Furnaces"