Frozen: Freezers for Your Lab Samples
Laboratory Freezers
Laboratory Freezers
When your samples require storage at low temperatures, a laboratory freezer is a necessary investment. General-purpose laboratory freezers store your samples safely while ultra-low temperature freezers are best for more critical samples. Maintain sample security with key locking doors or access codes.
When looking for a freezer that’s best for your lab, consider these options:
  • Glass doors to maintain full view of samples at all times
  • Optional chart recorders for continuous visible monitoring of sample temperatures
  • Sealing to minimize moisture accumulation
  • Space-saving designs, such as compact/undercounter models if your lab space is tight
  • Touch-screen control panels to control settings and authority levels
  • Built-in alarm to alert you to temperature rise or power loss
  • On-board computer to store up to 15 years worth of temperature and event data
Your selection of options will depend on your budget and your specific requirements. Some units are Energy Star rated for maximum energy conservation.
Laboratory freezer accessories to enhance your space include storage racks and boxes, remote alarm systems, and more.
View our selection of laboratory freezers.
Be the first to comment on "Frozen: Freezers for Your Lab Samples"