2 Types of High-Temperature Vacuum Ovens
StableTemp® Electronic-controlled High-Temperature Vacuum Oven
StableTemp® Electronic-controlled High-Temperature Vacuum Oven
Vacuum ovens can dry samples without the oxidation or residue that may result with other ovens. High-temperature vacuum ovens offer precise, uniform control of temperature. Simple, safe operation is the hallmark of these two types of high-temperature vacuum ovens:
  • Electronic-Controlled Oven offers wraparound heating elements to ensure even heating. An electronic control back-up prevents overheating. The front panel controls include primary temperature control failure indicators, purge gas and vacuum control valves, vacuum gauge, and temperature control. Gas and vacuum system connectors on the rear panel let you replace air with inert gas. Glance through the window to view temperature on the dial thermometer inside the chamber.
  • Microprocessor-Controlled Oven provides self-diagnostic indicators, a back-up thermostat, and dual controls to prevent damage to samples. Program it for up to eight on/off cycles per day. The front panel indicators alert you to open thermocouple, open heater, or overtemperature conditions.
View all laboratory ovens.
Be the first to comment on "2 Types of High-Temperature Vacuum Ovens"