How to Analyze Protein and Moisture Content Anywhere
Cole-Parmer® Portable NIR Food Analyzer Kit
Cole-Parmer® Portable NIR Food Analyzer Kit
To ensure the best volume of protein, oil, and moisture in dairy and wheat products (and secure your profitability), check it with a food analyzer. These analyzers or meters help you with maintain food quality by determining the content of your products, usually in less than one minute.
The meter shown analyzes protein and oil both on an “as-is” basis and a constant moisture (CM) basis. Because it is portable, you can take it with you on the plant floor or in the field.
How does the food analyzer work? Simply pour your sample of about 100 grams into the holder and place the holder into the meter to scan. The results are quickly revealed and you can download the data to your 486 computer or better with the included calibration software and RS-232 cable (USB).
View our selection of food analyzers.
Be the first to comment on "How to Analyze Protein and Moisture Content Anywhere"