The Swiss-engineered Masterflex Ismatec pumps are renowned for their clocklike precision and manufactured according to strict ISO 9001:2008 standards. These independent channel control (ICC) pumps provide lower pulsation, better reproducibility, and longer tubing life. Variable flow and bidirectional capabilities in each channel add to their flexibility. The uniqueness of the ICC allows for individual channel calibration.
A much-desired feature—computer compatibility—enables you to monitor the pumps remotely, make adjustments, and gather and analyze measurement data.
Masterflex Ismatec pumps are ideal for:
- Dissolution testing in pharmaceutical dosing
- Chromatography applications
- Adding a reagent to a reactor
- R&D
Select from 2, 3, 4, 6, 8, and 12 roller options. Single-channel Masterflex Ismatec Reglo Quick pumps offer quick tubing changes while Flowmaster® pumps are designed for heavy-duty processes and increased hygienic requirements.
Be the first to comment on "High Precision, Low Flow? Look for Masterflex Ismatec Pumps"