Maintaining Water Quality: the Clean Water Rule
Water Quality
Polluted water impairs the health of individuals, endangers wildlife, and increases water treatment costs. About 44% of assessed streams and 64% of assessed lake acres are not clean enough to allow fishing and swimming, according to the Environmental Protection Agency (EPA).Pollutants may include any number of chemicals and nutrients, but the leading violators include bacteria, mercury, phosphorus, nitrogen, and organic material.
While water quality was the impetus behind the Clean Water Act, legalized in 1972, the Act was unclear when it came to protecting many US waterways. The Clean Water Rule, finalized in May 2015, protects streams and wetlands scientifically shown to have the greatest impact on downstream water quality and which form the foundation of US water sources.
Measuring Water Quality
Water quality is seldom a constant in any body of water. When it rains, contaminants such as oil, grease, chemicals from motor vehicles, pesticides, road salts, and more run off streets untreated into rivers, lakes, and bays. Nitrate runoff from fertilizer can concentrate in drinking water if not removed by water treatment.
Water scientists test water using equipment designed to detect these contaminants. Measurement instruments used in the field and in the laboratory include:
  • pH Meters reveal pH level, which can change significantly if chemicals, acid rain or drainage have contaminated the water
  • Dissolved Oxygen Meters monitor water pollution by testing oxygen availability. Low levels of dissolved oxygen may indicate the presence of organic material decomposition
  • Turbidity Meters assess water clarity and particulates in the water
  • Total Dissolved Solids (TDS) Meters measure anything that cannot be seen, including minerals, salts, metals, and more
  • Multiparameter Meters test a number of parameters, which may include pH, conductivity, dissolved oxygen, temperature, turbidity, ion concentrations, barometric pressure, and salinity
  • Colorimeters determine the concentration of a large range of contaminants in a water sample
View our selection of water quality instruments.
Be the first to comment on "Maintaining Water Quality: the Clean Water Rule"