Save time in the digestion process with polypropylene disposable digestion cups. These cups act as digestion vessels, graduated cylinders, and storage containers—no need to use separate graduated cylinders in your transfer process.
Disposable digestion cups are approved for hot plate digestions. They are certified for low metals background and volume to 0.5%—and EPA approved for digestions.
Clarified homopolymer polypropylene construction offers greater chemical resistance and heat tolerance than commonly used co-polymer polypropylene vials. With screw-top caps with a polyethylene foam liner—only metal-free polypropylene and polyethylene come in contact with your sample.
If you’re using the HotBlock® Digestion System or HotBlock® Pro Blocks Systems, these cups will fit perfectly in your system.
Have questions? Give our technical experts a call. View our selection of disposable digestion cups.
Be the first to comment on "Eliminate Transfer Steps with Disposable Digestion Cups"