Timed Right: Timers for Labs
Cole-Parmer® Triple-Display Three-Channel Digital Timer
Cole-Parmer® Triple-Display Three-Channel Digital Timer
Timing is everything and certainly when you need to program time intervals in the lab or kitchen. The Three-Channel Digital Timer certainly made an impact on an Academic/Professional researcher from Iowa. This customer told us:
“This timer does everything: Times three separate situations at once; it’s a stop watch, clock and alarm clock. This is my second Cole-Palmer timer and I use it for both professional and personal applications. The first one was purchased in 1998 (14 years ago) and used daily. I needed to replace it, not because it wore out, but because I dropped it on a very hard surface! Truly a great product. To say I was thrilled to find that the exact timer was still available is an understatement. I’ve tried less expensive multiple line timers … and they just don’t compare.”
Why did this timer get a five-star rating? Its three timer channels can be used in any combination of up/down timing. You can start all three channels at once or individually; distinct audible alarm sounds and “TIMES UP” flashes on the display when the set time is reached. The memory function returns the display to the programmed time.
Essentially, it’s easy, inexpensive, and does the job!
Be the first to comment on "Timed Right: Timers for Labs"