Is your peristaltic pump equipped with open-head sensor interlock? If it is, you are one step ahead of the curve and providing an extra layer of protection for your operators. If not, you should read the article Enhance Workplace Safety with Open-Head Sensor Interlock in authored by Gregg Johnson, our global senior product manager for the Masterflex® and Masterflex Ismatec® product lines for over 19 years.
In the article, Gregg explains how peristaltic pumps work and how equipping them with open-head sensor interlock adds an extra layer of protection for operators who are tasked with tubing changes or maintaining the pumps. He also explains how it can be beneficial for peristaltic pumps operating in a system where programmable logic controller (PLC) or supervisory control and data acquisition (SCADA) systems control the pumps.
Open-head sensor interlock is available on Masterflex Easy-Load pump heads compatible with all L/S® and I/P® drives. A retrofit solution is also available to upgrade existing Masterflex peristaltic pumps. The retrofit system is comprised of a control box and a new open-head interlock sensor-equipped pump head.
Flow Control is online and in print to serve industries where fluid handling is a requirement.
Be the first to comment on "Masterflex Open-Head Sensor Interlock Featured in Flow Control"