No need for gateways or adapters with our new Masterflex® Advanced Networking Drives with options for popular networking protocols. These communication protocols cover a broad range of connectivity and communication with most advanced automation control systems. These drives have protocols that cover a broad range of connectivity and communication with most advanced automation control systems. The drives are ideal for industrial networking, bioprocessing, plant automation, or water/wastewater applications.
The new drive options include native Ethernet/IP and Profibus bi-directional communication with Bluetooth Low Energy (BLE) connectivity and a downloadable iOS-based mobile app to permit local pump monitoring and control. A Cloud-based remote pump monitoring capability is available to provide operational status and alerts. The drives are also compatible with most advanced automation control systems, such as Allen-Bradley®, Siemens®, Emerson®, and Honeywell®.
Advanced networking drives with Ethernet I/P are available for Masterflex L/S® and I/P® drives in either a benchtop console or a process- suitable NEMA 4X (IP66) enclosure. Drives are also equipped with Masterflex open-head interlock sensor technology.
Be the first to comment on "New Masterflex® Advanced Networking Drives"