If you’ve been using both a lab mixer and an industrial mixer for your single application because you can’t get what you need using just one type of mixer, check out this digital batch mixer.
With one of the highest torque settings on the market, the Cole-Parmer® Digital Batch Mixer has enough power to mix up to 52 gallons (200 L) at a maximum viscosity of 180,000 cps, yet provides a smooth and quiet operation. This batch mixer is ideal for laboratory and industrial uses in basic pre-mix, complex pilot scale, or full production batches.
This portable mixer provides user-friendly operation with easy impeller adjustment, PLC/PC programmability, data logging, and intuitive graphic touch-screen control. Selection of time and speed can be completed with a gloved hand or pencil tip. RS-232 and USB interfaces to record date, time, speed, and torque data.
Other functions include alarms, timers, torque zero, torque Min/Max setting, and clockwise and counterclockwise rotation. Safety features include a shaft/check guard, chemical-resistant housing, and overload, overheat and vibration protection. A safety screen protects the display from splashes. Mounting bolts make installing on existing equipment easy. Optional accessories allow for stand or drum installation and optimal shaft angle in the vessel.
For more information on the digital batch mixer, contact our product experts at 1-800-323-4340.
Be the first to comment on "New Digital Batch Mixer is Ideal for Lab and Industrial Use"