Three Minute DNA or RNA Extraction and Free Arcis Sample Prep Kit Promotion
Acris Sample Preparation Kit
Here’s an easy way for you to save hours in the lab. You no longer have to wait 40 to 90 minutes to extract DNA or RNA. Use the Arcis DNA Sample Prep Kit for rapid extraction of either DNA or RNA in three minutes with high yields. This process is ideal for any molecular biologist who wants to reduce the time in which DNA and RNA samples can be obtained.
Arcis DNA Sample Prep Kit Takes Two Simple Steps
Existing sample preparation kits currently require laborious washing, heating, and centrifugation steps that typically take 40 to 90 minutes to extract DNA or RNA from a sample. The Arcis DNA Sample Prep Kit requires two simple steps that are completed in three minutes, to provide high-quality, PCR-ready templates with no need for additional laboratory equipment such as centrifuges or hot blocks. The Arcis Prep Kit is ideal for applications such as PCR, qPCR, isothermal amplification (LAMP), sequencing, genetic profiling (electrophoresis) and forensic studies. The extraction system can also be easily integrated into automated sample processing or liquid handling systems.
The Arcis DNA Prep Kit, if used for extractions from fluids like blood, requires as little as 30 µL of sample as a starting material. The kit is also scalable in accordance with stated ratios in the prep kit manual and unlike other products, the prep kit can extract both DNA and RNA. This dual purpose removes the need for separate DNA and RNA extraction kits and, combined with the unrivaled three-minute sample processing time, saves time and money for labs of all sizes.
 FREE Arcis Prep Kit Promotion
The Arcis DNA sample prep kit is ideal for use with our PCRmax PCR and qPCR hardware, so for a limited time, we are offering FREE Arcis Kit(s) with a qualifying purchase of a PCRmax thermal cycler.
Purchase a PCRmax Alpha Cycler or Eco 48 Real-Time qPCR System, you will get the following:
See details. Hurry! Offer ends October 31, 2017. SHOP NOW
Be the first to comment on "Three Minute DNA or RNA Extraction and Free Arcis Sample Prep Kit Promotion"