Q: Is your refrigerator running?
A: Yes.
Q: Better go catch it!
This classic telephone prank may be the oldest joke in the book, but it’s no joke for many of you who rely on a laboratory refrigerator to keep your samples or vaccines safely stored. If your laboratory refrigerator is running, but you aren’t keeping a close eye on the temperature, you are taking a chance on losing your valuable samples or vaccines. It’s important to monitor your refrigerator temperature. So, what can you do?
Invest in the following:
- Stand-alone laboratory refrigerator(s) with enough space to accommodate your maximum inventory without crowding
- Stand-alone lab freezer(s) with enough space to accommodate your maximum inventory without crowding
- Digital data logger (DDL) with remote monitoring for every laboratory refrigerator or lab freezer you own, plus a backup in the event one malfunctions.
Digital data loggers with remote monitoring help you stay connected to your critical environments—wherever you go. Data loggers, such as the Digi-Sense Traceable Data Logging refrigerator/freezer thermometers with TraceableLIVE, help you connect to your data via smartphone, tablet, or PC using wireless capability. You can view current temperature conditions anywhere and with a cloud-based interface, you can send email, text, and push notifications. Plus, data loggers with NIST-traceable calibration ensure accuracy.
What are the major considerations in choosing digital data logger? Learn more in TraceableLive: Connected Lab Considerations
Learn how digital data loggers meet requirements for the vaccine for kids program.
Browse our complete line of data loggers.
Be the first to comment on "Is Your Laboratory Refrigerator Running… at the Right Temperature?"