How Does a Refractometer Work?
Refractometer
New Cole-Parmer® Digital Refractometer
What is a refractometer?
A refractometer is an instrument used for measuring concentrations of aqueous solutions. It requires only a few drops of liquid and is used as lab equipment throughout the food, agricultural, chemical, and manufacturing industries. A refractometer can be found in a mechanic’s toolbox, a winery, and even a gemologist’s desk. Home beer brewers and aquarium keepers also use it. Refractometers can test everything from battery acid to the authenticity of emeralds. This handy device is most often used in the food and beverage industry.
How does a refractometer work?
When light enters a liquid it changes direction; this is called refraction. Refractometers measure the degree to which the light changes direction, called the angle of refraction. A refractometer takes the refraction angles and correlates them to refractive index (nD) values that have been established. Using these values, you can determine the concentrations of solutions. For example, solutions have different refractive indexes depending on their concentration.
The prism in the refractometer has a greater refractive index than the solution. Measurements are read at the point where the prism and solution meet. With a low concentration solution, the refractive index of the prism is much greater than that of the sample, creating a large refraction angle and a low reading. The reverse would happen with a high concentration solution.
Browse the new line of Cole-Parmer digital refractometers.
The % Brix Scale
The % Brix Scale shows the concentration percentage of the soluble solids content of a sample (water solution). The soluble solids content is the total of all the solids dissolved in the water, beginning with sugar, salts, protein, acids, etc.; the measurement reading value is the value of the sum total of those. The Brix scale is calibrated to the number of grams of cane sugar contained in 100 g of water. When measuring a sugar solution, the % Brix reading should perfectly match the actual concentration.
Sample fluid
% Brix
Cutting oils
0 to 8
Oranges
4 to 13
Carbonated beverages
5 to 15
Apples
11 to 18
Grapes and wines
14 to 19
Concentrated juices
42 to 68
Condensed milk
52 to 68
Jams and jellies
60 to 70
Common Refractive Indexes: Refractive index readings are temperature-dependent.
Sample fluid
Temperature
Refractive index
Methanol
25°C
1.326
Acetone
25°C
1.357
Ethanol
25°C
1.359
Acetic acid
25°C
1.370
Benzene
25°C
1.498
Paraffin oil
20°C
1.412
Palm oil
20°C
1.456
Olive oil
20°C
1.471
Methyl salicylate
25°C
1.522
Methyl iodide
25°C
1.740
Learn more about refractometers.
Browse our complete line of laboratory equipment.
Be the first to comment on "How Does a Refractometer Work?"