We’re Talking OEM Capabilities at MD&M East
OEM pumpsMasterflex Medical OEM Pump
Stop and see us in booth #1471
We are exhibiting at MD&M East from June 12 to 14, 2018! If you are at the show, stop by booth 1471 and talk to us about your OEM peristaltic pump projects. With our extensive experience in OEM design capabilities for peristaltic pumps, we are ready to assist you.
Also, ask about our new Masterflex® Medical OEM Pump specifically designed to address the needs of medical device companies that require a reliable and easy-to-use pump component for medical devices used for ablation, surgical irrigation, liposuction, endoscopy, and angioscopy therapies. The clean design and quiet operation make this pump ideal for use in surgical areas where the noise level is a concern.
Pump Features
The pump features flow rates up to 300 mL/min and pressure more than 4 bar (60 psig). Various tubing configurations allow for a flexible fluid path including side to side, bottom entry or exit, or a combination of side/bottom entry or exit. Easy access to the pump chamber allows operators to perform fast tubing changes. A stainless steel cover with viewing window adds strength and the ability to confirm pump operation. An integrated open-head sensor stops the drive when the pump head is opened.
Learn how to create a custom peristaltic pump.
Browse our complete line of Masterflex pumps
Be the first to comment on "We’re Talking OEM Capabilities at MD&M East"