A simple way to track conditions of items on the GO!
The Digi-Sense Temperature Data Logger with TraceableGO™ Wireless Capability and Calibration allows you to easily track conditions of reagents, clinical samples, vaccines, pharmaceuticals, and food items during transport. Simply connect to data via any Bluetooth®-enabled device using the downloadable Bluetooth app. Configure data loggers from your device using the TraceableGO app to set parameters and alarms. Once the shipment is delivered, enable Bluetooth to export data to your device and immediately see if shipment contents were exposed to conditions outside the set parameters. The data loggers compact design can fit into almost anything—even a small cooler. Plus, get accurate, reliable measurements with included NIST-traceable calibration.
Use the TraceableLive® app and upload data to the cloud
You can also subscribe to cloud-based TraceableLive service to upload your data to the cloud—ideal for tracking data from multiple loggers or to comply with regulatory agency requirements. This low-cost subscription provides unlimited trip data to be saved on the cloud and the ability to send a report to an email.
Browse our complete line of data loggers.
Be the first to comment on "How To Track Conditions of Items During Transport"