What is a desiccator?
What is a desiccator
Learn more about the desiccator
Cole-Parmer DC-400 Automatic Desiccator Cabinet with N2 Purge Port, One Shelf; 50 L
A sealable canister, or cabinet, known as desiccator keep contents dry (and dust-free) with the help of desiccant. These economical storage containers are used for electronics, reagents, optical equipment, electronic parts, PCB boards, reagents and other humidity-sensitive items and are sealed to absorb moisture. Once a moisture-sensitive object is placed inside the desiccator with the desiccant (or drying agent), the object will remain dry or at a set humidity level below ambient on adjustable automatic cabinets. These canisters or cabinets are sometimes referred to as desiccant dryers or desiccant dehumidifiers. Desiccators are important tools for scientists who perform chemical reactions because the units help them ensure accurate chemical measurements before starting a chemical reaction. Desiccators also are used to transport and store samples for short- or long-term.
Read the entire article Learn more about desiccators
Need a desiccator?
Choose from vacuum, nonvacuum or cabinet. Here are a few to consider:
Cole-Parmer® Essentials Vacuum Desiccators with White Base, PP/PC
vacuum desiccatorThese vacuum desiccators can hold a vacuum up to 29” (740 mm) Hg for 24 hours without any greasing. The top dome with molded-in knurled knob is constructed of rigid, transparent polycarbonate (PC), providing you a clear view of your sample.
Cole-Parmer® Essentials Glass Dome Desiccators
Manufactured from durable borosilicate glass, these clear glass dome desiccators are designed for analytical work. The borosilicate glass lid features a molded-in knob for easy handling of the cover. To create an airtight seal, it is recommended you grease the rim and lid of the desiccator.
Cole-Parmer® DC-400 Automatic Desiccator Cabinets
Keeping a constant RH level has never been easier. The electronic dehumidification system in this desiccator cabinet includes a permanent desiccant and a hygrometer to keep the relative humidity level between 25 and 55%. The desiccant automatically recycles by discharging absorbed moisture through an external shutter.
See all desiccators
Related Articles
Which Chamber is Best for Your Lab? (Part 1: Desiccators)
Sign up for the blog banner
Be the first to comment on "What is a desiccator?"