Reduce cross-contamination and increase accuracy in weighing with clean glassware
Good laboratory technique requires clean glassware, labware and weighing dishes, because the most carefully executed piece of work may give an erroneous result if the dirty glassware is used. When weighing in particular, the accuracy can be thrown off if even a fingerprint is on the glass. In all instances, glassware must be physically and chemically clean, and in some cases, it needs to be bacteriologically clean or sterile.
Glassware must be kept grease-free
The safest criteria of cleanliness is uniform wetting of the surface by distilled water. This is especially important in glassware used for measuring the volume of liquids. Grease and other contaminating materials will prevent the glass from becoming uniformly wetted. This will alter the volume of residue adhering to the walls of the glass container and thus affect the volume of liquid delivered. Furthermore, in pipettes and burettes, the meniscus will be distorted and the correct adjustments cannot be made. The presence of small amounts of impurities may also alter the meniscus.
Wash labware as soon as possible after use
If a thorough cleaning is not possible immediately, put glassware to soak in water. If labware is not cleaned immediately, it may become impossible to remove the residue.
Learn more. Read the article, Caring for Your Glassware.
Need glassware
Source: Corning Corporation
Be the first to comment on "How to Clean Lab Glassware"