How to Navigate Pharma-Grade Certifications
An illustration depicting a collaborative compliance process. Multiple hands are shown around a table, pointing to various items like a certificate of compliance, a laptop, financial documents, and a calculator. The background features a blueprint-style world map with green leaves, symbolizing sustainability and global coordination.
Why Certifications Matter in Pharmaceutical Development and Manufacturing
Pharmaceutical laboratories and manufacturing facilities operate under strict oversight from regulatory bodies like the U.S. Food and Drug Administration (FDA), the European Medicines Agency (EMA), and others. These organizations establish guidelines to ensure the safety and quality of medicines and healthcare technologies. Certifications act as a stamp of approval, confirming that materials and components meet rigorous safety and quality standards.
In this article you’ll learn
  • Key Certifications in the Pharmaceutical Industry
  • The Role of Documentation
  • How to Obtain Documentation
Key Certifications for Fluid Handling Components in the Pharmaceutical Industry
1. United States Pharmacopeia (USP) Class VI
What is USP Class VI?
The United States Pharmacopeia (USP) sets standards for the quality, purity, strength, and consistency of drugs and their components. USP Class VI, one of six classifications for plastics, is the most stringent. It certifies that plastic materials do not cause harmful reactions or long-term effects when they come into contact with living tissues. To earn this designation, materials must pass three biocompatibility tests: acute systemic toxicity, intracutaneous toxicity, and implantation. These tests ensure that materials used in pharmaceutical production are safe and do not leach harmful substances.
Why Use It?
USP Class VI is vital for drug manufacturing due to its strict biocompatibility and low-toxicity requirements. It ensures that no harmful chemicals leach into products, making it essential for fluid-contact surfaces like tubing, fittings, and single-use systems.
A close-up image of a hand wearing a blue disposable glove adjusting a stainless steel sanitary fitting. The fitting features a clamp and a barbed connection, commonly used in laboratory or industrial fluid transfer systems.
Products that meet USP Class VI standards:
2. U.S. Food and Drug Administration (FDA) CFR 21
What is FDA CFR 21?
The U.S. Food and Drug Administration (FDA) plays a critical role in regulating pharmaceutical products, ensuring their safety, efficacy, and quality. The FDA’s Code of Federal Regulations (CFR) Title 21 is the set of regulations that governs food and drug safety in the United States.
Two common FDA certifications used in the pharmaceutical industry are:
  • FDA CFR 21 177.2600: Governs rubber materials used in repeated contact with food, ensuring they don’t introduce harmful substances.
  • FDA 21 CFR 177.1520: Specifies the types of olefin polymers, including polyethylene and polypropylene, that are safe for use in food-contact applications.
Why Use It?
While originally designed for materials in contact with food, it is widely applied in the pharmaceutical industry. They ensure that elastomeric materials used in tubing, gaskets, and seals meet strict safety standards.
Gloved hand holding flexible tubing in a laboratory setting, with a peristaltic pump and lab equipment, including flasks and test tubes, in the background.
FDA compliant fluid handling components:
3. European Pharmacopoeia (EP) 3.2.9
What is EP 3.2.9?
For companies selling pharmaceuticals in Europe, adhering to the European Pharmacopoeia (EP) is essential. EP 3.2.9 is a standard that establishes guidelines for evaluating elastomeric closures. This guideline ensures component materials are free from harmful extractables and leachables, which could compromise product quality.
Why use it?
This guideline emphasizes the importance of comprehensive testing to mitigate risks associated with product contamination and ensure that pharmaceutical products maintain their quality throughout their shelf life. It is crucial for pharmaceutical manufacturers seeking to market their products in Europe, as it demonstrates adherence to high-quality standards and regulatory requirements.
Gloved hands holding clear braided tubing on a stainless steel surface, highlighting its use in cleanroom environments.
Sani-Tech® STHT®-80 premium silicone tubing with high burst pressure capability.
European Pharmacopoeia (EP) 3.2.9 compliant products:
4. BSE/TSE Free Certification
What is BSE and TSE?
BSE (Bovine Spongiform Encephalopathy) and TSE (Transmissible Spongiform Encephalopathy) are neurodegenerative diseases that can be transmitted through animal-derived materials. It is critical to ensure that the products used throughout the development and manufacturing process do not use any animal-derived ingredients, and the product does not come into contact with animal products during storage or transportation.
Why use it?
BSE/TSE Free Certification confirms that the materials used are free from any animal-derived substances that could carry the risk of BSE or TSE transmission. This certification is often required for materials used in single-use systems, bioprocessing, and sterile drug manufacturing to ensure the highest levels of safety and compliance. Although the prevalence of TSEs is low globally, it’s important to take precautions to minimize their spread.
Scientist in cleanroom attire handling AseptiQuik connectors, demonstrating sterile connection of tubing components with precision.
Products that are TSE and BSE free:
The Role of Documentation
While certifications ensure the quality and safety of fluid handling components, comprehensive documentation plays an equally vital role in regulatory compliance. Proper documentation helps establish traceability and accountability, which is crucial for audits, regulatory inspections, and maintaining quality standards.
Types of Documentation:
  • Certificates of Conformance (CoC): Documents that certify a product meets specified requirements, giving manufacturers confidence in the materials they use. A Certificate of Compliance typically includes several critical pieces of information to ensure full traceability and confidence in the product, including but not limited to the description, lot number, and shelf life where applicable.
  • Validation Packets: Compressive packets that provide the necessary regulatory, performance, and safety standards for the testing and validation of component materials. Validation packets may include applicable compliance certificates, extractables and leachables, biocompatibility reports, or performance testing.
How to Obtain Documentation
Cole-Parmer compliance certificate for fluid handling products with sample product details, showcasing a free on-demand lookup for certificates of compliance.
Sample Compliance Document
Working with reputable suppliers like Cole-Parmer, who are well-versed in documentation and regulatory requirements, can simplify the process of obtaining the necessary certifications and documentation.
Cole-Parmer offers a convenient self-service certificate of compliance lookup function that allows you to quickly access Certificates of Conformance (CoCs) for your purchased products. This feature requires no login—simply enter the product lot number to retrieve the necessary documentation, streamlining the process of ensuring compliance.
In addition to CoCs, Cole-Parmer offers material validation packets for applicable fluid handling components. Contact a Cole-Parmer representative to inquire about validation packets available for fluid handling products. https://www.coleparmer.com/fluid-handling-solutions
Warning: These products are not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Related Articles/Blog Posts
https://www.coleparmer.com/blog/fda-compliance-temperature-monitoring-traceablelive/
https://www.coleparmer.com/blog/how-does-traceablelive-support-21-cfr-part-11-compliance/
https://www.coleparmer.com/blog/lab-accreditation/
https://www.coleparmer.com/tech-article/regulatory-agencies-and-approvals
%d bloggers like this: