Availability
Control Type
IP Rating
Max Flow Rate (mL/min)
Max Flow Rate Per Channel (mL/min)
Max RPM
Max RPM (rpm)
Min Flow Rate (mL/min)
Low Flow Rate Peristaltic Pumps
Custom cut tubing
CUSTOM CUT TUBING
Low flow peristaltic pumps are ideal for autoanalysis, flow cell, sipper, OEM, and low flow feed and transfer applications. They offer compact size, simple controls, and the low-maintenance and cost-effectiveness of peristaltic pumps. When selecting a pump, choose a system that offers a flow rate that's 50-75% of your systems peak capabilities to get a longer tube life as well as more flexibility.