Pump Series (1)
Number of Channels
Max Flow Rate (mL/min)
Min Flow Rate (mL/min)
Product Type
Brand
Price Range
Availability
Masterflex L/S Pump Heads
High Accuracy, Repeatable Performance. Deliver flow rates from 0.0006 to 3400 mL/min. Pump heads are designed to work best with Masterflex L/S precision pump tubing, ensuring confidence in pump performance. Most pump designs accept continuous lengths of tubing, allowing you to: Customize tubing lengths to your application. Simplify sanitary setups—no breaks in tubing.Reduce setup/cleanup time. Eliminate difficult fittings and clamps. Increase tubing life—periodically stop the pump, move tubing 15 to 20 cm (6 to 8 in.), reload, and continue pumping.
APPLY FILTERS | 2
filter byclear all
24 RESULTS
SORT BY
Showing 1 - 12 of 24
2>