Availability
Control Type
Max Flow Rate (LPM)
Max Flow Rate (mL/min)
Max Flow Rate Per Channel (mL/min)
Max RPM
Min Flow Rate (mL/min)
Min Flow Rate Per Channel (mL/min)
Medium Flow Rate Peristaltic Pumps
Masterflex Single Use
Custom Disposable Sterile Bioprocess Assemblies.
Custom cut tubing
CUSTOM CUT TUBING
Medium flow peristaltic pumps combine good flow and flexibility and are ideal for autoanalysis, flow cell, sipper, OEM, and medium flow feed and transfer applications. When selecting a pump, choose a system that offers a flow rate that's 50-75% of your systems peak capabilities to get a longer tube life as well as more flexibility.
APPLY FILTERS | 1
filter byclear all
12 RESULTS
SORT BY
Showing 1 - 12 of 12