Harvesting Process Workflow
Centrifuge
Centrifuges are instruments that use centrifugal force to separate colloidal particles or substances of different densities, to remove moisture, for microfiltration, or to simulate gravitational effects. When selecting a centrifuge consider the following: anticorrosive materials, compact footprint, refrigeration, rotor versatility, and throughput.
View Centrifuge Products
Buffer
Buffers are solutions of known pH used to stabilize the acidity of a liquid product. It is imperative to maintain control of the pH of a product to avoid cell death, avoid drug degradation, or improve the efficacy of delivery.
View Buffer Products
Washing Tank
Tanks, vessels, and bottles store product between the drug production stages. They are sterilized or involve a sterilized barrier between the vessel and the product (such as a tank liner) to mitigate the risk of contamination. Tank ports are pre-ordered to fit the needs of the application and sample bottles include a vent filter to avoid pressurizing the bottle during dispensation and to avoid cross-contamination.
View Washing Tank Products
Micro Filtration
Depending on the production floor or laboratory throughput, the filter will range in capacity and act as a rough filter in the microscale.
View Micro Filtration Products
Washing Tank
Tanks, vessels, and bottles store product between the drug production stages. They are sterilized or involve a sterilized barrier between the vessel and the product (such as a tank liner) to mitigate the risk of contamination. Tank ports are pre-ordered to fit the needs of the application and sample bottles include a vent filter to avoid pressurizing the bottle during dispensation and to avoid cross-contamination.
View Washing Tank Products
Buffer
Buffers are solutions of known pH used to stabilize the acidity of a liquid product. It is imperative to maintain control of the pH of a product to avoid cell death, avoid drug degradation, or improve the efficacy of delivery.
View Buffer Products
Ultra-Filtration
One of the final steps in the harvest workflow, ultrafiltration acts to remove the finest impurities from the target macromolecule product resulting a concentrate. This is usually done at a high pressure.
View Ultra-Filtration Products
Washing Tank
Tanks, vessels, and bottles store product between the drug production stages. They are sterilized or involve a sterilized barrier between the vessel and the product (such as a tank liner) to mitigate the risk of contamination. Tank ports are pre-ordered to fit the needs of the application and sample bottles include a vent filter to avoid pressurizing the bottle during dispensation and to avoid cross-contamination.
View Storage Vessel Products
Centrifuge
Centrifuges are instruments that use centrifugal force to separate colloidal particles or substances of different densities, to remove moisture, for microfiltration, or to simulate gravitational effects. When selecting a centrifuge consider the following: anticorrosive materials, compact footprint, refrigeration, rotor versatility, and throughput.
View Centrifuge Products
Buffer
Buffers are solutions of known pH used to stabilize the acidity of a liquid product. It is imperative to maintain control of the pH of a product to avoid cell death, avoid drug degradation, or improve the efficacy of delivery.
View Buffer Products
Washing Tank
Tanks, vessels, and bottles store product between the drug production stages. They are sterilized or involve a sterilized barrier between the vessel and the product (such as a tank liner) to mitigate the risk of contamination. Tank ports are pre-ordered to fit the needs of the application and sample bottles include a vent filter to avoid pressurizing the bottle during dispensation and to avoid cross-contamination.
View Washing Tank Products
Micro Filtration
Depending on the production floor or laboratory throughput, the filter will range in capacity and act as a rough filter in the microscale.
View Micro Filtration Products
Ultra-Filtration
One of the final steps in the harvest workflow, ultrafiltration acts to remove the finest impurities from the target macromolecule product resulting a concentrate. This is usually done at a high pressure.
View Ultra-Filtration Products