3-ply Ear Loop Face Mask with Melt-Blown Non-woven Fabric, >98% BFE (2.5 µm), >93% PFE (0.5 µm); Box of 50
Representative image only.
Mfr # MASK1-3P – Item # EW-78901-21
Three layers of fabric provide better protection
  • Flat, pleated design is easy to put on and take off
  • Comfortable to wear—soft ear loops and adjustable nose cover
  • Bacterial filtration efficiency is ≥95%
  • Manufactured in an FDA registered facility
$21.00USD / Box of 50
BUY MORE & SAVE
  • 1-4$21.00USD / Box of 50
  • 5+$18.90USD / Box of 50
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene(PP)
  • ColorBlue
  • Description3-ply Ear Loop Face Mask with Melt-Blown Non-woven Fabric, >98% BFE (2.5 µm), >93% PFE (0.5 µm); Box of 50
Key Features
  • Flat, pleated design is easy to put on and take off
  • Comfortable to wear—soft ear loops and adjustable nose cover
  • Bacterial filtration efficiency is ≥95%
  • Manufactured in an FDA registered facility
More About this Item
Disposable 3-ply face mask with soft ear loops and an adjustable nose cover is designed to be lightweight for comfort. Three layers include a polypropylene outer layer, nonwoven melt-blown filter layer, and polypropylene inner layer. Nonsterile.