304 SS mixing shaft, 10 mm dia x 500 mm L

Representative image only.
Item # EW-04370-02

$98.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Shaft MaterialStainless Steel
  • Shaft Diameter (in)0.375
  • Shaft Diameter (mm)10
  • Shaft Length (in)19.6
  • Shaft Length (mm)500
  • Description304 SS mixing shaft, 10 mm dia x 500 mm L

More About this Item

Mixing shaft for use with any 10 mm dia or adjustable 3/8" dia chuck. Measures 10 mm dia x 500 mm L. Use only with propellers listed separately.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat