3D Instruments 75514-21B55 Digital Test Gauge, Vac To 30 Psi

Representative image only.
3D Instruments Mfr # 75514-21B55 – Item # EW-68299-02
Outstanding accuracy, resolution, and stability
  • NEMA 4 rated case
  • LCD features bargraph and backlight
  • ±0.05% full scale accuracy for pressure and ±0.25% for vacuum
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Accuracy±0.05% full-scale (pressure); ±0.25% full-scale (vacuum)
  • Dial Size (in)4.5
  • Dial Size (mm)114
  • Min Pressure (PSI)-5
  • Max Pressure (PSI)30
  • Min Temperature (° F)14
  • Max Temperature (° F)131
  • Min Temperature (° C)-10
  • Max Temperature (° C)55
  • Wetted Materials316 SS
  • Lens MaterialPolycarbonate
  • Process Connection1/4\" NPT(M)
  • Unit Of Measurementpsi
  • Display5.5-digit LCD, 0.65\"H with 20-segment bar graph
  • Battery TypeThree AA (included)
  • Battery Life2000 hours
  • OutputRS-232
  • Enclosure RatingNEMA 4
  • Media CompatibilityGases and liquids compatible with wetted materials
  • Resolution0.001 psi
  • DescriptionDigital Test Gauge, Vac To 30 Psi
  • Warranty1 year
1 Year WarrantyCanadian Standards Association

Key Features

  • NEMA 4 rated case
  • LCD features bargraph and backlight
  • ±0.05% full scale accuracy for pressure and ±0.25% for vacuum
  • 1 year warranty

More About this Item

Ideal for calibrating pressure transmitters, switches, and gauges. Gauge provides numerous user-configurable features such as adjustable sample rates, dampening, tare, custom engineering units, Min/Max, and password-protected field calibration. Gauge is built to comply with requirements for Class 1, Division 1, Groups A, B, C, and D hazardous locations. Large, backlit 5 1/2 digit display has 0.65" high digits as well as a 20 segment bar graph. Gauge is intrinsically safe for use in Class 1, Division 1, Groups A, B, C, and D hazardous locations.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat