3M 3500 Personal Air Monitor, Organic Vapors, 10/bx
Representative image only.
3M Mfr # 3500 – Item # EW-40116-00
Simple and effective — badge-style design ensures easy compliance with air monitoring reulations.
$333.20USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Gases MeasuredOrganic vapors
  • Analysis IncludedNo
  • DescriptionPersonal Air Monitor, Organic Vapors, 10/bx
More About this Item
Monitors contain a sorbent wafer that collects airborne contaminants for later analysis. Clip unit to your clothing, wear for the duration of the sampling period, and send it to 3M® or any other certified gas chromatography lab to determine exposure.
Organic vapor monitor has a charcoal sorbent wafer that absorbs over 125 vapors— ideal for personal monitoring in a variety of settings.