This item is now discontinued. Please browse similar items
3M 9211 Three-Panel Particulate respirator with exhalation valve, N95, Box of 10
Representative image only.
3M Mfr # 9211 – Item # EW-86401-10
Lightweight respirator delivers comfort, convenience, and security
  • Improves visibility, prevents collapsing, and fits many face sizes
  • Individually wrapped to prevent contamination during storage
SPECS & DESCRIPTION
Specifications & Description
  • RatingN95
  • ComplianceNIOSH
  • ColorWhite
  • 3M Item Number9211
  • DescriptionThree-Panel Particulate respirator with exhalation valve, N95, Box of 10
Key Features
  • Improves visibility, prevents collapsing, and fits many face sizes
  • Individually wrapped to prevent contamination during storage
More About this Item
Low profile makes these respirators ideal for use with other safety equipment. The soft inner panel feels comfortable to the skin. The adjustable nose clip provides a custom fit and a long-lasting, secure seal.
Use in textile, dry chemical, surface preparation, food preparation, woodworking, milling, mineral processing, and other industries that create dusty environments. Respirator can also be used in environments with liquid or non-oil based sprays that do not contain harmful vapors.
Approval: NIOSH
Model 86401-10 features a one-way exhalation valve that reduces build-up of moisture and allows comfortable breathing.