3M BE-12-3 Belt-Mounted PAPR Respirator Hood, 3/Pk
Representative image only.
3M Mfr # BE-12-3 – Item # EW-40220-11
High efficiency filters for extremely fine dust hazards
  • Up to eight hours operation on a single charge
SPECS & DESCRIPTION
Specifications & Description
  • ComplianceNIOSH
  • DescriptionBelt-Mounted PAPR Respirator Hood, 3/Pk
Key Features
  • Up to eight hours operation on a single charge
More About this Item
This positive pressure hooded respirator is ideal for workers with facial hair who can not wear tight fitting mask. It is an excellent choice in applications where there is very fine dust, such as in asbestos abatement. Assembled components comprise a NIOSH approved respirator. Order components separately.
The standard hood features a chin shroud with elastic band that helps maintain a tight face fit. The extended bib hood can be tucked into workers coveralls allowing a flow of air to help cool the user. Both style hoods are made from Tychem® for excellent particulate and liquid protection.
The breathing tube brings air from blower assembly to the hood.
Blower assembly includes blower, HEPA filter with 99.97% efficiency to 0.3 micron, battery (charger sold separately), waist belt, and airflow indicator.