This item is now discontinued. Please browse similar items
3M DMS Data Management Software
Representative image only.
3M Mfr # DMS – Item # EW-50536-39
Quickly and accurately measure noise levels for exposure evaluation
  • Variable, user-selectable data logging intervals with user-determined measurement parameters
  • Store numerous noise surveys in memory for download
  • Large backlit LCD
  • IP65-rated design with ergonomic soft grip
  • Multilingual user interface
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionData Management Software
Includes
Microphone windscreen, USB cable, carrying case, and lithium battery with charger.
Key Features
  • Variable, user-selectable data logging intervals with user-determined measurement parameters
  • Store numerous noise surveys in memory for download
  • Large backlit LCD
  • IP65-rated design with ergonomic soft grip
  • Multilingual user interface
  • Rechargeable lithium battery
More About this Item
These sound meters are engineered to accurately measure noise levels in highly variable environments. Instruments compute the average sound pressure level (Leq/Lavg) over the run time, helping you to more accurately assess occupational and environmental noise levels. The Detection Management Software (DMS) lets you retrieve, download, share and save instrument data, generate charts and reports, export and share recorded data, perform advanced calculations, and easily export data to a Microsoft® Excel® spreadsheet. The optional printer allows the immediate printing of measurement results.