9170 VIBRATING TACHOMETER
Representative image only.
Mfr # 9170 – Item # EW-98805-95
  • Compact size tachometers have smaller and fewer reeds
  • Ideal for tool case or pocket use
  • Popular in the textile industry for measuring speeds of sewing machines
PRODUCT OPTIONS
$229.40USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Min RPM Range (rpm)9000
  • Max RPM Range (rpm)13000
  • Display TypeAnalog
Key Features
  • Compact size tachometers have smaller and fewer reeds
  • Ideal for tool case or pocket use
  • Popular in the textile industry for measuring speeds of sewing machines
More About this Item
Accurate - Rugged - Dependable
Because these tachometers measure speeds by contact, this type of tachometer is the solution to many speed measuring problems, especially checking speeds of totally enclosed rotating equipment. These tachometers operate on the well known and time tested principle of resonance. The instrument is simply held against motor, turbine, pump, compressor, or other rotating equipment and the speed is shown by the vibration of a steel reed which is tuned to a certain standard speed.
Ruggedly constructed, permanent accuracy, simple to use. The instrument imposes no load and cannot be damaged by overspeeding. Accuracy is 0.5% of rated speed.
Other typical applications are outboard motors, electric shavers, centrifuges, etc.