A&D Weighing GXA-14 Density Determination Kit for Apollo Precision Analytical Balance
Representative image only.
A&D Weighing Mfr # GXA-14 – Item # EW-11109-48
Use your balance to determine density with these convenient kits
  • Expand the capabilities of your balance with these convenient kits
  • Ensure the kit corresponds to your correct A&D Weighing balance
  • Suitable for 0.0001 g readability models only
  • 5 year warranty
PRODUCT OPTIONS
$473.10USD / Each
$570.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDensity Determination Kit for Apollo Precision Analytical Balance
  • Warranty5 Years
5 Year Warranty
Key Features
  • Expand the capabilities of your balance with these convenient kits
  • Ensure the kit corresponds to your correct A&D Weighing balance
  • Suitable for 0.0001 g readability models only
  • 5 year warranty
More About this Item
These density determination kits, when combined with one of the A&D Weighing electronic balances, allows the density of a solid and liquid to be measured easily. Balances sold separately.