A&D Weighing HT-300CL Compact Scale for Packing and Checkweighing 310 x 0.1 g , Inc Carry Case
Representative image only.
A&D Weighing Mfr # HT-300CL – Item # EW-11116-31
Compact & portable scale with multiple weighing units
  • Battery or AC adapter operation
  • LCD display
  • Multiple weighing units
  • Storage/carrying case included
PRODUCT OPTIONS
$174.30USD / Each
$210.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
Includes
4 AA alkaline batteries, storage/carrying case, 2 year warranty
Key Features
  • Battery or AC adapter operation
  • LCD display
  • Multiple weighing units
  • Storage/carrying case included
More About this Item
The HT-Series compact scales operate on 4 AA alkaline batteries or the optional 120VAC adapter. The handy storage and carrying case also accommodates storage for batteries. The HT-Series feature a low battery indicator, overload indication and a stable indicator for convenience.