Accuform KCC617 Portable Group Slot Lockout Lock Box, Red; 10" x 6" x 4-1/4"
Representative image only.
Accuform Mfr # KCC617 – Item # EW-40504-24
Ideal when multiple personnel are working on various lockout machinery and equipment
$205.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPortable Group Slot Lockout Lock Box, Red; 10" x 6" x 4-1/4"
More About this Item
Keys of locked-out devices are kept in the lock box, which is controlled by a foreman or group manager. Once the job is completed and workers are away from the equipment, the foreman unlocks the box so the workers can reclaim access to the keys.
Thirteen slot holes are located on the top of the lock box to allow approved workers to secure a padlock, ensuring that no one has access to the keys until all the padlocks are removed. Steel lockbox features a carrying handle for easy portability.
Call Us At1-847-549-7600
Live chat