Accuform KDD121 Single-Pole Circuit Breaker Lockout, Red; Breaker Toggles Up To 9/16"
Representative image only.
Accuform Mfr # KDD121 – Item # EW-40504-04
Lock out electrical power at the source – right at the circuit breaker, no need to lock the entire panel
  • Manufactured of rigid plastic in visible bright red
  • No modifications required, attach directly to the breaker toggle and panel to prevent the breaker from being switched on once padlock is in place
  • Fits over most major brands of breaker toggles, in the off position
  • Manufactured of rigid plastic in visible bright red
  • Accommodates padlocks with shackle diameter up to 9/32" (7.1 mm) (sold separately)
  • Single-pole style fits over breaker toggle up to 9/16" (14.3 mm) width with a recessed hole
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSingle-Pole Circuit Breaker Lockout, Red; Breaker Toggles Up To 9/16\"
Key Features
  • Manufactured of rigid plastic in visible bright red
  • No modifications required, attach directly to the breaker toggle and panel to prevent the breaker from being switched on once padlock is in place
  • Fits over most major brands of breaker toggles, in the off position
  • Manufactured of rigid plastic in visible bright red
  • Accommodates padlocks with shackle diameter up to 9/32" (7.1 mm) (sold separately)
  • Single-pole style fits over breaker toggle up to 9/16" (14.3 mm) width with a recessed hole
More About this Item
No need to lock out the entire panel; these compact, universally designed lockouts fit over most major brands of breaker toggles in the "off" position. Accommodates padlocks (not included) with shackle diameter up to 9/32" (7.1 mm).
Single-Pole Style fits over breaker toggle up to 9/16" (1.4 cm) width with a recessed hole.